Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis C Virus Core Antigen Antibody (C7-50), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1202740IR
Description
Hepatitis C Virus Core Antigen Monoclonal specifically detects Hepatitis C Virus Core Antigen in Human, Mouse, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence.Specifications
Hepatitis C Virus Core Antigen | |
Monoclonal | |
DyLight 755 | |
Core protein p19, HCcAg, HCV core antigen, HCV core protein | |
Mouse | |
Protein A or G purified | |
RUO | |
951475 | |
Human, Mouse, Virus | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
C7-50 | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
POLY | |
Purified HCV core-GST fusion protein (genotype 1b). | |
0.1 mL | |
Primary | |
Detects hepatitis C virus (HCV) core protein from transfected human and primate cell lines. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction