Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis C Virus Core Antigen Antibody (C7-50), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1202740JF646
Description
Hepatitis C Virus Core Antigen Monoclonal specifically detects Hepatitis C Virus Core Antigen in Human, Mouse, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence.Specifications
| HepatitisCVirusCoreAntigen | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| POLY | |
| Purified HCV core-GST fusion protein (genotype 1b). | |
| 0.1 mL | |
| Primary | |
| Detects hepatitis C virus (HCV) core protein from transfected human and primate cell lines. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| C7-50 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence | |
| Core protein p19, HCcAg, HCV core antigen, HCV core protein | |
| Mouse | |
| Affinity Purified | |
| Virology Bacteria and Parasites | |
| 951475 | |
| Human, Mouse, Virus | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction