Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis C Virus Core Antigen Antibody (C7-50), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1202740MFV610
Description
Hepatitis C Virus Core Antigen Monoclonal antibody specifically detects Hepatitis C Virus Core Antigen in Human, Mouse, Virus samples. It is validated for Flow CytometrySpecifications
Hepatitis C Virus Core Antigen | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse, Virus | |
Purified |
Flow Cytometry | |
C7-50 | |
Flow Cytometry | |
Core protein p19, HCcAg, HCV core antigen, HCV core protein | |
Purified HCV core-GST fusion protein (genotype 1b). | |
0.1 mL | |
Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
951475 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction