Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis C Virus Core Antigen Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093581MFV610
Description
Hepatitis C Virus Core Antigen Polyclonal antibody specifically detects Hepatitis C Virus Core Antigen in Human samples. It is validated for Western BlotSpecifications
Hepatitis C Virus Core Antigen | |
Polyclonal | |
Western Blot | |
Core protein p19, HCcAg, HCV core antigen, HCV core protein | |
Hepatitis C Virus (HCV) core protein (HCV-C1) | |
0.1 mL | |
Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
951475 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction