Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis C Virus Core Antigen Monoclonal Antibody (6A1), FITC
GREENER_CHOICE

Catalog No. MA190321
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA190321 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA190321 Supplier Invitrogen™ Supplier No. MA190321
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-90321 detects Hepatitis C core antigen from virus samples.

HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis C Virus Core Antigen
Applications ELISA, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone 6A1
Concentration 1 mg/mL
Conjugate FITC
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene Alias HCV core; HCVgp1; HepC; HepC core; polyprotein
Host Species Mouse
Immunogen Recombinant protein.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.