Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis C Virus Core Antigen Antibody (C7-50), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1202740APCCY7
Description
Hepatitis C Virus Core Antigen Monoclonal specifically detects Hepatitis C Virus Core Antigen in Human, Mouse, Virus samples. It is validated for Flow Cytometry.Specifications
Hepatitis C Virus Core Antigen | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
POLY | |
Purified HCV core-GST fusion protein (genotype 1b). | |
0.1 mL | |
Primary | |
Detects hepatitis C virus (HCV) core protein from transfected human and primate cell lines. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
C7-50 | |
Flow Cytometry | |
Core protein p19, HCcAg, HCV core antigen, HCV core protein | |
Mouse | |
Affinity Purified | |
Virology Bacteria and Parasites | |
951475.0 | |
Human, Mouse, Virus | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction