Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis C Virus Core Antigen NS3/NS4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA128927
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA128927 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA128927 Supplier Invitrogen™ Supplier No. PA128927
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis C Virus Core Antigen NS3/NS4
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias HCV core; HepC; HepC core
Host Species Goat
Immunogen Recombinant (E. coli) polypeptide containing core and NS3 and NS4 regions.
Purification Method IgG fraction
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.