Learn More
Invitrogen™ Hepatitis C Virus Core Antigen Polyclonal Antibody

Description
The PA1-73106 antibody reacts with Hepatitis C virus core antigen. Potential applications for PA1-73106 are numerous including ELISA, Western blot and Immunohistochemical applications. In addition , this product may be used in place of neat serum in almost any appropriate antibody-based technique. The PA1-73106 antibody was raised against recombinant core (genotype 1a).
Specifications
Specifications
| Antigen | Hepatitis C Virus Core Antigen |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | HCV core; HCVgp1; HepC; HepC core; polyprotein |
| Host Species | Goat |
| Immunogen | Recombinant core (genotype 1a). |
| Purification Method | IgG fraction |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.