Learn More
Description
Specifications
Specifications
| Antigen | Hepatitis C Virus E2 |
| Applications | Immunocytochemistry, ELISA, Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 |
| Formulation | PBS with 0.09% Sodium Azide |
| Host Species | Goat |
| Immunogen | Recombinant Hepatitis C Virus E2 (genotype 1a) |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
