Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis C Virus E2 Antibody - BSA Free, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10062654
Description
Hepatitis C Virus E2 Polyclonal specifically detects Hepatitis C Virus E2 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Hepatitis C Virus E2 | |
Polyclonal | |
Unconjugated | |
PBS with 0.09% Sodium Azide | |
Goat | |
Affinity Purified | |
RUO | |
This product is specific for the Hepatitis C Virus E2 antigen. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000 | |
HCV E4, HEPC | |
Recombinant Hepatitis C Virus E2 (genotype 1a) | |
0.5 mL | |
Primary | |
Virus | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction