Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis C Virus NS3 Monoclonal Antibody (24-8)
GREENER_CHOICE

Catalog No. PIMA121375
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA121375 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA121375 Supplier Invitrogen™ Supplier No. MA121375
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-21375 detects Hepatitis C Virus in Virus samples.

Hepatitis C virus (HCV) is a small (55-65 nm in size), enveloped, positive-sense single-stranded RNA virus of the family Flaviviridae. The hepatitis C virus is the cause of hepatitis C and some cancers such as liver cancer (hepatocellular carcinoma, abbreviated HCC) and lymphomas in humans. Nonstructural protein 3 (NS3), also known as p-70, is a viral nonstructural protein that is 70 kDa cleavage product of the hepatitis C virus polyprotein. It acts as a serine protease. C-terminal two thirds of the protein also acts as helicase and nucleoside triphosphatase. First (N-terminal) 180 aminoacids of NS3 has additional role as cofactor domains for NS2 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis C Virus NS3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 24-8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.2
Gene Alias HCV; HCV NS3; Hepatitis C Virus; HepC; HepC NS3
Host Species Mouse
Immunogen A highly antigenic polypeptide consisting of essential sequences of at least 60 residues in length, which were selected from genes encoding the NS-3 region of Chinese HCV strains.
Purification Method Protein G
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.