Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis C Virus NS3 Monoclonal Antibody (C44D)
GREENER_CHOICE

Catalog No. PIMA17345
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17345 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17345 Supplier Invitrogen™ Supplier No. MA17345
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7345 antibody reacts with Hepatitis C virus NS3 in viral and infected tissue samples. MA1-7345 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7345 antibody was raised against NS3 protein.

Hepatitis C virus (HCV) is a small (55-65 nm in size), enveloped, positive-sense single-stranded RNA virus of the family Flaviviridae. The hepatitis C virus is the cause of hepatitis C and some cancers such as liver cancer (hepatocellular carcinoma, abbreviated HCC) and lymphomas in humans. Nonstructural protein 3 (NS3), also known as p-70, is a viral nonstructural protein that is 70 kDa cleavage product of the hepatitis C virus polyprotein. It acts as a serine protease. C-terminal two thirds of the protein also acts as helicase and nucleoside triphosphatase. First (N-terminal) 180 aminoacids of NS3 has additional role as cofactor domains for NS2 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis C Virus NS3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone C44D
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias HCV; HCV NS3; Hepatitis C Virus; HepC; HepC NS3
Host Species Mouse
Immunogen NS3 protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.