Learn More
Invitrogen™ Hepatitis C Virus NS3 Monoclonal Antibody (F39C)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA17355
Description
The MA1-7355 antibody reacts with Hepatitis C virus NS3 in viral and infected tissue samples. This antibody is reactive towards genotypes 1a, 1b, and 2a. MA1-7355 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7355 antibody was raised against HCV NS3 protein.
Hepatitis C virus (HCV) is a small (55-65 nm in size), enveloped, positive-sense single-stranded RNA virus of the family Flaviviridae. The hepatitis C virus is the cause of hepatitis C and some cancers such as liver cancer (hepatocellular carcinoma, abbreviated HCC) and lymphomas in humans. Nonstructural protein 3 (NS3), also known as p-70, is a viral nonstructural protein that is 70 kDa cleavage product of the hepatitis C virus polyprotein. It acts as a serine protease. C-terminal two thirds of the protein also acts as helicase and nucleoside triphosphatase. First (N-terminal) 180 aminoacids of NS3 has additional role as cofactor domains for NS2 protein.
Specifications
Hepatitis C Virus NS3 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
ELISA, Western Blot, Immunocytochemistry | |
F39C | |
Unconjugated | |
HCV; HCV NS3; Hepatitis C Virus; HepC; HepC NS3 | |
NS3 protein. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.