Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis C Virus NS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5111976
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5111976 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5111976 Supplier Invitrogen™ Supplier No. PA5111976
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hepatitis C Virus NS3 Polyclonal Antibody for Western Blot

Hepatitis C virus (HCV) is a small (55-65 nm in size), enveloped, positive-sense single-stranded RNA virus of the family Flaviviridae. The hepatitis C virus is the cause of hepatitis C and some cancers such as liver cancer (hepatocellular carcinoma, abbreviated HCC) and lymphomas in humans. Nonstructural protein 3 (NS3), also known as p-70, is a viral nonstructural protein that is 70 kDa cleavage product of the hepatitis C virus polyprotein. It acts as a serine protease. C-terminal two thirds of the protein also acts as helicase and nucleoside triphosphatase. First (N-terminal) 180 aminoacids of NS3 has additional role as cofactor domains for NS2 protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis C Virus NS3
Applications Western Blot
Classification Polyclonal
Concentration 0.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias HCV; HCV NS3; Hepatitis C Virus; HepC; HepC NS3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of Hepatitis C virus NS3 protein (HCV 2a (strain JFH-1)). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.