Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis C Virus NS5 Monoclonal Antibody (BGN/1246/5G7)
GREENER_CHOICE

Catalog No. PIMA182923
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA182923 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA182923 Supplier Invitrogen™ Supplier No. MA182923
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In Western blot, this antibody detects a major band of ~50 kDa with minor bands at ~35, ~40, and ~67 kDa. This reflects bands found in DTT reduced blots fo the immunizing antigen and in immunoblots using a known antibody-positive serum.

HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis C Virus NS5
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone BGN/1246/5G7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene Alias HCV; HCV NS5; Hepatitis C Virus; HepC; HepC NS5; NICE-3
Host Species Mouse
Immunogen Synthetic peptide corresponding to the NS-5 antigen region of the native protein.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.