Learn More
Invitrogen™ Hepatitis C Virus NS5 Polyclonal Antibody

Description
The PA1-73109 antibody reacts with Hepatitis C virus NS5 antigen. PA1-73109 has been successfully used in ELISA, and Western blot applications. The PA1-73109 antibody was raised against recombinant NS5 (genotype 1a). Recommended short term storage, less than 6 months, is at 2-8ºC. For longer term storage, aliquot and freeze.
Specifications
Specifications
| Antigen | Hepatitis C Virus NS5 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 4-5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | HCV; HCV NS5; Hepatitis C Virus; HepC; HepC NS5; NICE-3 |
| Host Species | Goat |
| Immunogen | Recombinant NS5 (genotype 1a). |
| Purification Method | IgG fraction |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.