Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C Virus NS5a Monoclonal Antibody (J63I)
GREENER_CHOICE

Catalog No. PIMA17344
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17344 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17344 Supplier Thermo Scientific Supplier No. MA17344
Add to Cart
Add to Cart

Hepatitis C Virus NS5a Monoclonal Antibody (J63I)

Hepatitis C Virus NS5a Monoclonal Antibody for ICC/IF, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis C Virus NS5a
Applications ELISA, Immunocytochemistry
Classification Monoclonal
Clone J63I
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias HCV; HCV NS5; Hepatitis C Virus; HepC; HepC NS5; NICE-3
Host Species Mouse
Immunogen NS5a protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.