Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis C Virus NS5b Monoclonal Antibody (H09B)
GREENER_CHOICE

Catalog No. MA17342
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA17342 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA17342 Supplier Invitrogen™ Supplier No. MA17342
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The MA1-7342 antibody reacts with Hepatitis C virus NS5b. This antibody detects genotype 1a and 1b, but not 2a. MA1-7342 has been successfully used in ELISA, Immunofluorescence and Western blot applications. The MA1-7342 antibody was raised against NS5b protein.

HCV is a positive, single-stranded RNA virus in the Flaviviridae family. The genome is approximately 10,000 nucleotides and encodes a single polyprotein of about 3,000 amino acids. The polyprotein is processed by host cell and viral proteases into three major structural proteins and several non-structural protein necessary for viral replication. Several different genotypes of HCV with slightly different genomic sequences have since been identified that correlate with differences in response to treatment with interferon alpha.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis C Virus NS5b
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone H09B
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene Alias HCV; HCV NS5; Hepatitis C Virus; HepC; HepC NS5; NICE-3
Host Species Mouse
Immunogen NS5b protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.