Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis D Virus Large delta Protein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117451
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117451 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117451 Supplier Invitrogen™ Supplier No. PA5117451
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Centrifuge briefly prior to opening.

HDV is the only virus in the genus, Deltaviridae. HDV is not classified into a viral family because it is a unique virus dependent on HBV. HDV is a co-infection of HBV. The envelope of HDV particles contains the Hepatitis B surface antigen (HBsAg). The production and transmission of HDV is entirely dependent on HBV to provide HBsAg. Thus, HDV is considered a satellite virus of HBV. Unlike a classical satellite virus, however, HDV does not share sequence similarity with HBV, and it can replicate independently of HBV. The nucleocapsid is made up of 60 large and small delta antigens. These are the only proteins encoded by HDV.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis D Virus Large delta Protein
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.34 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias HDV
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Hepatitis D virus Large delta protein (HDV genotype I (strain D380)). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.