Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis D virus Recombinant Rabbit Monoclonal Antibody (HL1055)
GREENER_CHOICE

Catalog No. PIMA542361
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542361 Supplier Invitrogen™ Supplier No. MA542361
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Hepatitis D virus (HDV), also known as hepatitis delta virus, is a defective RNA virus that requires the hepatitis B virus (HBV) for its replication and assembly. Discovered in 1977, HDV is dependent on the HBV surface antigen (HBsAg) for completing its life cycle, as it does not encode its own envelope proteins. HDV infection leads to a severe form of viral hepatitis, often resulting in more rapid progression to liver cirrhosis, liver failure, and hepatocellular carcinoma compared to HBV infection alone. HDV primarily co-infects or super-infects individuals with HBV, impacting approximately 12-72 million people worldwide. New therapeutic approaches are being explored to manage this difficult-to-treat disease, given its association with adverse liver outcomes. Chronic HDV/HBV co-infection has a higher incidence of severe liver disease within a short period, typically within 5-10 years.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis D virus
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1055
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias HDV; Hepatitis Delta Virus
Host Species Rabbit
Immunogen Full length Hepatitis D virus Large data and Small delta recombinant protein (Hepatitis delta virus genotype I (isolate D380)).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.