Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis E Virus Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Hepatitis E Virus |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017931
![]() |
Novus Biologicals
NB10093583SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10093583
![]() |
Novus Biologicals
NB10093583 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Hepatitis E Virus Polyclonal specifically detects Hepatitis E Virus in Human samples. It is validated for Western Blot.Specifications
Hepatitis E Virus | |
Western Blot 1:100-1:2000 | |
Polyclonal | |
Rabbit | |
Human | |
HEV | |
IgG | |
Affinity Purified | |
Reacts with residues 627-637 [CPECRTLGLQG] of the 71 kDa HEV capsid protein. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
PBS with 0.02% Sodium Azide | |
residues 627-637 [CPECRTLGLQG] of the 71 kDa HEV capsid protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title