Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis E Virus Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10093583PCP
Description
Hepatitis E Virus Polyclonal antibody specifically detects Hepatitis E Virus in Human samples. It is validated for Western BlotSpecifications
Hepatitis E Virus | |
Polyclonal | |
Western Blot | |
HEV | |
residues 627-637 [CPECRTLGLQG] of the 71 kDa HEV capsid protein. | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction