Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis E Virus Capsid Mouse anti-Virus, Clone: EB2.2.G10.B4.A10, Novus Biologicals™

Mouse Monoclonal Antibody
$515.50 - $1262.00
Specifications
Antigen | Hepatitis E Virus Capsid |
---|---|
Clone | EB2.2.G10.B4.A10 |
Concentration | 1 mg/mL |
Dilution | ELISA |
Applications | ELISA |
Description
Hepatitis E Virus Capsid Monoclonal antibody specifically detects Hepatitis E Virus Capsid in Virus samples. It is validated for ELISASpecifications
Hepatitis E Virus Capsid | |
1 mg/mL | |
ELISA | |
Unconjugated | |
Mouse | |
Virus | |
ORF2, pORF2, Secreted protein ORF2 | |
IgG1 κ | |
Protein G purified | |
Antibody |
EB2.2.G10.B4.A10 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Phosphate Buffered Saline, pH 7.4, Filter Sterile | |
HEV ORF2 aa 1-660 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title