Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis E Virus Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA522822
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA522822 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA522822 Supplier Invitrogen™ Supplier No. PA522822
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specificity: Reacts with residues 627-637 [CPECRTLGLQG] of the 71 kDa HEV capsid protein. Immunogen sequence: CPECRTLGLQ G.

Hepatitis E virus (HEV) is a leading cause of acute viral hepatitis, mainly transmitted via the fecal-oral route, often through contaminated water. HEV is a positive-sense single-stranded RNA virus belonging to the Hepeviridae family. Human HEV infections are primarily caused by genotypes 1 and 2, which are restricted to humans, while genotypes 3 and 4 are zoonotic and also infect various animals such as pigs and deer. HEV is known for its broad host range and potential to cause epidemics, leading to significant morbidity and mortality, especially in pregnant women. Recent studies have highlighted the extrahepatic manifestations of HEV, including neurological, renal, and hematological complications. HEV infections often result in self-limited acute hepatitis; however, chronic HEV infections can occur in immunocompromised individuals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis E Virus
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias HepE; HEV
Host Species Rabbit
Immunogen residues 627-637 of the 71 kDa HEV capsid protein.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.