Learn More
Invitrogen™ Hepatitis E Virus Polyclonal Antibody

Description
PA1-4101 detects the hepatitis E virus (HEV) variant identified in the United States, designated HEV US-u3-2e. PA1-4101 has been successfully used in Western blot procedures. The PA1-4101 immunogen is a synthetic peptide corresponding to residues C(627) P E C R T L G L Q G(637) of the HEV capsid protein.
Specifications
Specifications
| Antigen | Hepatitis E Virus |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | HepE; HEV |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to residues C(627) P E C R T L G L Q G(637) of the HEV capsid protein. |
| Purification Method | Antigen affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.