Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis G Virus E2 Monoclonal Antibody (UIE2-1)
GREENER_CHOICE

Catalog No. PIMA518088
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518088 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518088 Supplier Invitrogen™ Supplier No. MA518088
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody recognizes a linear epitope in the GBV-C virus glycoprotein E2.

Hepatitis G virus is from a group of viruses called GB viruses (GB virus A, GB virus B and GB virus C) and is also designated as GB virus C (GBV C). HGV is a positive stranded RNA virus, with a genome of 10 KB and appears, on the basis of genome length and organisation, to be a member of the Flaviviridae and distinct from the hepatitis C virus (HCV). HGV infection appears to be transmitted following receipt of blood from HGV RNA positive donors, however, these patients have only moderate liver enzyme elevations and do not develop chronic liver disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis G Virus E2
Applications ELISA, Western Blot
Classification Monoclonal
Clone UIE2-1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Alias GBC glycoprotein E2; GBV-C; HepG; HGV
Host Species Mouse
Immunogen GBV-C glycoprotein E2.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.