Learn More
Invitrogen™ Hepatitis G Virus Polyclonal Antibody

Description
This antibody is reactive against Hepatitis G Virus / Hepatitis GBV-C in Human samples. Immunogen sequence: CDKCEARQE.
Specifications
Specifications
| Antigen | Hepatitis G Virus |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene Alias | GBC glycoprotein E2; GBV-C; HepG; HGV |
| Host Species | Rabbit |
| Immunogen | Reacts with residues 2268-2276 of HGV. |
| Purification Method | Antigen affinity chromatography |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.