Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatitis G Virus Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14100 Supplier Invitrogen™ Supplier No. PA14100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4100 detects the hepatitis G virus (HGV). PA1-4100 has been successfully used in Western blot procedures. The PA1-4100 immunogen is a synthetic peptide corresponding to residues C(2268) D K C E A R Q E(2276) of HGV.

Hepatitis G virus is from a group of viruses called GB viruses (GB virus A, GB virus B and GB virus C) and is also designated as GB virus C (GBV C). HGV is a positive stranded RNA virus, with a genome of 10 KB and appears, on the basis of genome length and organisation, to be a member of the Flaviviridae and distinct from the hepatitis C virus (HCV). HGV infection appears to be transmitted following receipt of blood from HGV RNA positive donors, however, these patients have only moderate liver enzyme elevations and do not develop chronic liver disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatitis G Virus
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias GBC glycoprotein E2; GBV-C; HepG; HGV
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues C(2268) D K C E A R Q E(2276) of HGV.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.