Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis Virus A59 NSP9Mouse anti-Virus, Clone: 2C6.H1, Rockland™
SDP

Catalog No. 501990559
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-0559 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-0559 Supplier Rockland Immunochemicals Supplier No. 200301A56
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Directed against the MHV-A59 nsp9 protein. No cross reactivity occurs with SARS CoV nsp9. Cross reactivity with homologues from other sources has not been tested.

The nonstructural protein 9 (nsp9) is one of the Mouse hepatitis virus replicase cleavage products, encoded by ORF1a. Nsp9 is an RNA-binding protein that is thought to be part of the viral replication complex, which is associated with intracellular membranes.

Specifications

Antigen Hepatitis Virus A59 NSP9
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2C6.H1
Concentration 2.1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene ORF1a
Gene Accession No. P0C6V0
Gene Symbols ORF1a
Host Species Mouse
Immunogen This antibody was produced in mice by repeated immunizations with E.coli derived full-length MHV-A59 nsp9 protein. This protein is part of the viral replicase polyprotein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489749
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.