Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatocyte Growth Factor Ab-1 Mouse Monoclonal Antibody, Epredia™

Catalog No. MS651P1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
100 μL
200 μL
500 μL
1 mL
Concentration:
1mg/mL
200μg/mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
MS651P1 500 μL 200μg/mL
MS651P0 100 μL 200μg/mL
MS651P 1 mL 200μg/mL
MS651P1ABX 100 μL 1mg/mL
MS651PABX 200 μL 1mg/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MS651P1 Supplier Epredia Supplier No. MS651P1

Ensure accurate, reproducible results with Epredia™ Hepatocyte Growth Factor Ab-1, Mouse Monoclonal Antibody.

Hepatocyte Growth Factor (HGF), also known as Scatter Factor (SF) or Hepatopoietin A. Membrane receptor for HGF/SF is the product of the proto-oncogene c-met, a tyrosine kinase. HGF/SF is a pleiotropic growth factor which is produced by mesodermally derived cells, such as Kupffer cells/macrophages, endothelial cells, or hepatic fat-storing cells.

Host Species: Mouse

Clone: 4C12.1

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant human HGF protein

Molecular Weight: ∽82kDa

Recommended for:

  • Western Blotting (Not verified)

Note:

USA: RUO; Int'l: RUO

Specifications

Antigen Hepatocyte Growth Factor Ab-1
Classification Monoclonal
Clone 4C12.1
Concentration 200μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant human HGF protein
Quantity 500 μL
Regulatory Status RUO
Research Discipline Growth Factor
Primary or Secondary Primary
Target Species Human
Form Concentrated
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.