Learn More
Description
Host Species: Mouse
Clone: 4C12.1
Isotype: IgG2b
Species Reactivity: Human. Others not known.
Epitope: Not determined
Immunogen: Recombinant human HGF protein
Molecular Weight: ∽82kDa
Recommended for:
- Western Blotting (Not verified)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Hepatocyte Growth Factor Ab-1 |
| Classification | Monoclonal |
| Clone | 4C12.1 |
| Concentration | 1mg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Recombinant human HGF protein |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Research Discipline | Growth Factor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.