Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatocyte Specific Antigen Antibody (HepPar1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 4 publications
$269.00 - $512.00
Specifications
Antigen | Hepatocyte Specific Antigen |
---|---|
Clone | HepPar1 |
Concentration | 0.2mg/mL |
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24527200
![]() |
Novus Biologicals
NBP2452720.02MG |
0.02 mg |
Each for $269.00
|
|
|||||
NBP24527201
![]() |
Novus Biologicals
NBP2452720.1MG |
0.1 mg |
Each for $512.00
|
|
|||||
NBP24527202
![]() |
Novus Biologicals
NBP2452720.2MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ensure accurate, reproducible results in Immunohistochemistry (Paraffin), Immunofluorescence
Hepatocyte Specific Antigen Monoclonal specifically detects Hepatocyte Specific Antigen in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
Hepatocyte Specific Antigen | |
0.2mg/mL | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Mouse | |
Human, Mouse, Canine | |
Hepatocyte, HSA | |
IgG1 | |
Protein A or G purified | |
Hepatocyte Paraffin 1 or HepPar1 localizes to the mitochondria of hepatocytes. It is a sensitive marker for distinguishing hepatocellular carcinomas (HCC) from other metastatic carcinomas as well as cholangio-carcinomas. HCC s occur primarily in the stomach, but they are also found in many other organs. The Hepatocyte Specific Antigen may also be a useful marker for intestinal metaplasia. Reportedly, strong expression of the Hepatocyte Specific Antigen correlates with smaller tumor size and longer patient survival. Occasionally, Hepatocyte Specific Antigen is also found in gastric carcinomas as well as in a few other non-hepatic tumors. |
HepPar1 | |
Immunohistochemistry, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
Extract of a formalin-fixed, rejected-allograft of a human liver | |
Primary | |
Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title