Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatocyte Specific Antigen Antibody (HSA98), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234645AF405
Description
Hepatocyte Specific Antigen Monoclonal specifically detects Hepatocyte Specific Antigen in Human samples. It is validated for ELISA.Specifications
| Hepatocyte Specific Antigen | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Hepatocyte, HSA | |
| HEP-3B human hepatocellular carcinoma cells | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry, Immunofluorescence | |
| HSA98 | |
| Immunocytochemistry/Immunofluorescence | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Monoclonal antibodies (monoclonal antibodys). In frozen sections, it stains hepatic cells and may be used as a marker of the liver. Cell preparations of hepatocellular carcinoma biopsies or cell lines are found to bind HSA98 on the cell surface. This monoclonal antibody stains liver hepatocytes in frozen human liver sections and is positive on the cell surface of human liver carcinomas. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction