Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatocyte Specific Antigen Antibody (OCH1E5), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234699C
Description
Hepatocyte Specific Antigen Monoclonal specifically detects Hepatocyte Specific Antigen in Human, Canine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Hepatocyte Specific Antigen | |
Monoclonal | |
DyLight 650 | |
Hepatocyte, HSA | |
Extract of a formalin-fixed, rejected-allograft of a human liver | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
OCH1E5 | |
Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Canine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction