Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatocyte Specific Antigen Antibody (HepPar1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308970R
Description
Hepatocyte Specific Antigen Monoclonal antibody specifically detects Hepatocyte Specific Antigen in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ImmunofluorescenceSpecifications
Hepatocyte Specific Antigen | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Mouse, Canine | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
HepPar1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence | |
Hepatocyte, HSA | |
Extract of a formalin-fixed, rejected-allograft of a human liver | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction