Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Hepatocytes Monoclonal Antibody (OCH1E5)
GREENER_CHOICE

Catalog No. PIMA512417
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA512417 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA512417 Supplier Invitrogen™ Supplier No. MA512417
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-12417 targets Hepatocyte in IHC (P) applications and shows reactivity with Human samples. The MA5-12417 immunogen is formalin fixed failed human allograft liver that was mechanically disrupted.

Hepatocytes are the chief functional cells of the liver and perform a number of metabolic, endocrine and secretory functions. Roughly 80% of the mass of the liver is contributed by hepatocytes. In three dimensions, hepatocytes are arranged in plates that anastomose with one another.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Hepatocytes
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OCH1E5
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.09% sodium azide
Host Species Mouse
Immunogen Formalin fixed failed human allograft liver that was mechanically disrupted.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.