Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES, 0.5 M Buffer Solution, pH 7.6, PROMO, Thermo Scientific Chemicals

Catalog No. J61047AKPM
Click to view available options
Quantity:
250 mL

Applications

HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

Solubility

Fully miscible in water.

Notes

Stable under recommended storage conditions. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.3 g/mol
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 0.5 M
pH 7.6
Physical Form Liquid
Quantity 250 mL
Solubility Information Fully miscible in water.
Chemical Name or Material HEPES buffer solution

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.