Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals HEPES, 1.0 M Buffer Solution, pH 7.5, 250 mL PROMO

Catalog No. J60712AKPM
Click to view available options
:
250 mL

Applications

  • HEPES is common buffer for biological sciences, particularly used in cell culture to maintain physiological pH.
  • It acts a buffering component, which is used in the preparation of buffers.
  • It is described as one of the best all-purpose buffers available for use in biological research.

Solubility

  • Miscible with water.

Notes

  • Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30 g/mol
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 1.0 M
pH 7.5
Physical Form Liquid
Quantity 250 mL
Solubility Information Miscible with Water.
Chemical Name or Material HEPES buffer soln.
TSCA Yes
Recommended Storage Keep Cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.