Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES, 1M Solution, pH 7.3, Molecular Biology Grade, Ultrapure

Catalog No. AAJ16924AP
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene bottle

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
PubChem CID 23831
ChEBI CHEBI:42334
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

Concentration 1 M
pH 7.3
Physical Form Solution
Quantity 500 mL
Packaging Nalgene bottle
Beilstein 883043
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
Grade Molecular Biology
Chemical Name or Material HEPES
EINECSNumber 230-907-9
RTECSNumber TL6809000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.