Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEPES, Biological Buffer, 99%, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
5 kg
12 kg
Packaging:
Poly Bottle
Specifications
Specifications
CAS Max % | 1 |
For Use With (Application) | Pharma and Biopharma |
Physical Form | Crystalline |
Quantity | 500 g |
Loss on Drying | 0.005 |
Packaging | Poly Bottle |
Percent Purity | 99% |
Grade | Reagent |
Chemical Name or Material | 4-(2-Hydroxyethyl)-1-Piperazineethanesulfonic Acid |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction