Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES, Biological Buffer, 99%, Spectrum™ Chemical
SDP

Catalog No. 18610779
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
5 kg
12 kg
Packaging:
Poly Bottle

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

CAS Max % 1
For Use With (Application) Pharma and Biopharma
Physical Form Crystalline
Quantity 250 g
Loss on Drying 0.005
Packaging Poly Bottle
Percent Purity 99%
Grade Reagent
Chemical Name or Material 4-(2-Hydroxyethyl)-1-Piperazineethanesulfonic Acid

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.