Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEPES-Buffered Saline, 2X, pH 7.0, Low Endotoxin, PROMO, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mL
Specifications
Specifications
Chemical Name or Material | HEPES-buffered saline |
Color | Colorless |
Physical Form | Liquid |
Quantity | 250 mL |
RUO - Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction