Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES, cGMP Manufactured, 99.5%, Spectrum™ Chemical
SDP

Catalog No. p-7157568 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 kg
12 kg
25 kg

Catalog No. 18-603-104

Supplier: Spectrum Chemical Manufacturing Corporation H18041KGBL

Only null left
Add to Cart
Add to Cart

H1804, 7365-45-9, C8H18O4N2S

Spectrum™ Chemical HEPES, cGMP Manufactured is a zwitterionic biological buffer and is one of Good's buffers widely used in cell culture. Spectrum cGMP grade products that have been manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7365-45-9
Molecular Formula C8H18N2O4S
Molecular Weight (g/mol) 238.30
MDL Number MFCD00006158
InChI Key JKMHFZQWWAIEOD-UHFFFAOYSA-N
IUPAC Name 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
SMILES OCCN1CCN(CCS(O)(=O)=O)CC1

Specifications

CAS Max % 100%
For Use With (Application) Pharma and Biopharma
pH 5.4 to 5.6
Physical Form Crystalline Powder or Crystals
Quantity 1 kg
Packaging Poly Bottle
Percent Purity 99.5%
Grade Reagent
Chemical Name or Material 4-(2-Hydroxyethyl)-1-Piperazineethanesulfonic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.