Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES, Free Acid, Ultrapure Bioreagent, J.T.Baker™

Click to view available options
Quantity:
100g
500g
Packaging:
Poly bottle

Chemical Identifiers

7365-45-9
C8H18N2O4S
238.302
JKMHFZQWWAIEOD-UHFFFAOYSA-N
23831
CHEBI:42334
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
C1CN(CCN1CCO)CCS(=O)(=O)O

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Liquid Chromatography and Molecular Biology applications; For Laboratory,Research,or Manufacturing Use
Formula Weight 238.31
Packaging Poly bottle
Specific Gravity 1L = 0.00kg
Quantity 500g
Chemical Name or Material HEPES, Free Acid
Grade Ultrapure Bioreagent