Learn More
HEPES, non cGMP, Bioprocess Grade, Micro and Endotoxin Tested, bioCERTIFIED™, 1 kg, Spectrum Chemical

Description
100% Tested. 100% Transparent. 100% Trusted.
Spectrum Chemical bioCERTIFIED™ products are uniquely suited for bioprocessing by biopharmaceutical manufacturers. From drug discovery to pilot and scale-up to production, bioCERTIFIED™ products help simplify and standardize bioprocessing by ensuring consistent raw material quality. All bioCERTIFIED™ materials and finished goods are subject to in-house laboratory testing and analysis in cGMP-compliant facilities. This includes expanded testing for bioburden, endotoxin and elemental impurities.
- Extensive product breadth and packaging flexibility for upstream and downstream bioprocessing
- Expanded laboratory testing, certifications and documentation including additional lot testing for bioburden, endotoxins and elemental impurities
- Comprehensive regulatory and scientific documentation to help meet global quality, regulatory and compliance requirements
- Manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA-registered and inspected facilities
- Change control, lot traceability and supply chain transparency to eliminate unpredictability
Specifications
Specifications
Chemical Name or Material | 4-(2-Hydroxyethyl)-1-Piperazineethanesulfonic Acid |
CAS | 7365-45-9 |
CAS Max % | 100% |
For Use With (Application) | Molecular Biology |
Physical Form | Crystalline Powder |
Quantity | 1 kg |
Percent Purity | 99.5% |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.