Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Yttrium(III) nitrate hexahydrate, 99.9%, (trace metal basis)
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis

Specifications
Specifications
Color | White |
Physical Form | Adhering Crystalline Powder or Crystals |
Quantity | 100 g |
Assay Percent Range | 99.9% |
Linear Formula | Y(NO3)3·6H2O |
Merck Index | 14,10107 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol |
Formula Weight | 383.01 |
Percent Purity | 99.9% |
Grade | Trace Metal Basis |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement Harmful if swallowed. Causes serious eye irritation. Very toxic to aquatic life with long lasting effects. |
Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. Wash face,hands and any exposed skin thoroughly after handling. If eye irritation persists: Get medic |
RTECSNumber | ZG3750000 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction