Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hephaestin Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983787
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3787 Supplier Bioss Supplier No. BS15458R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

B1AJX8(HEPH) is similar to an iron transport protein found in mouse. The mouse protein is similar to ceruloplasmin, a serum multi-copper ferroxidase, and is thought to be a membrane-bound protein responsible for transport of dietary iron from epithelial cells of the intestinal lumen into the circulatory system. In mouse, defects in this gene can lead to severe microcytic anemia. Three transcript variants encoding different isoforms have been described for this gene. The protein encoded by this gene is similar to an iron transport protein found in mouse. The mouse protein is similar to ceruloplasmin, a serum multi-copper ferroxidase, and is thought to be a membrane-bound protein responsible for transport of dietary iron from epithelial cells of the intestinal lumen into the circulatory system. In mouse, defects in this gene can lead to severe microcytic anemia. Two transcript variants encoding different isoforms have been described for this gene.

Specifications

Antigen Hephaestin
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HEPH
Gene Accession No. Q9BQS7
Gene Alias C130006F04Rik; Cpl; haphaestin; Heph; Hephaestin; KIAA0698; mKIAA0698; sex linked anemia; sla; UNQ2562/PRO6242
Gene Symbols HEPH
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Hephaestin.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9843
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.