Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HEPN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565367
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565367 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565367 Supplier Invitrogen™ Supplier No. PA565367
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GLGIAPWVDGE SELEFRRLGM QGPLEALRRR ERNTQRASFS FSFLIALSPH TVDYCHSYEL FNRRWHGHVL ATQRPSLFI Highest antigen sequence identity to the following orthologs - mouse 27%, rat 27%.

This gene is expressed in the liver, and encodes a short peptide that is localized predominantly to the cytoplasm. Transient transfection studies showed that expression of this gene significantly inhibited cell growth, and it may have a role in apoptosis. Expression of this gene is downregulated or lost in hepatocellular carcinomas (HCC), suggesting that loss of this gene is involved in carcinogenesis of hepatocytes. Also to note is that this gene maps to the 3'-noncoding region of HEPACAM gene (GeneID:220296) on the antisense strand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HEPN1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HEPN1
Gene Accession No. Q6WQI6
Gene Alias cancer susceptibility gene HEPN1; HEPACAM opposite strand 1; hepatocellular carcinoma, down-regulated 1; HEPN1; Putative cancer susceptibility gene HEPN1 protein
Gene Symbols HEPN1
Host Species Rabbit
Immunogen Recombinant Human HEPN1. Recombinant protein control fragment (Product #RP-103488).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 641654
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.