Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HepPar-1 Monoclonal Antibody (OCH1E5), Zeta

Catalog No. 502215875
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-221-5875 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-221-5875 Supplier ZETA CORPORATION Supplier No. Z2087MS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody Cocktail

A recommended positive control tissue for this product is HCC, however positive controls are not limited to this tissue type. The primary antibody is intended for laboratory professional use in the detection of the corresponding protein in formalin-fixed, paraffin-embedded tissue stained in manual qualitative immunohistochemistry (IHC) testing. This antibody is intended to be used after the primary diagnosis of tumor has been made by conventional histopathology using non-immunological histochemical stains.

HepPar-1 Monoclonal antibody specifically detects HepPar-1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen HepPar-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OCH1E5
Concentration 200 μg/mL
Conjugate Unconjugated
Formulation tris with BSA, NP-40 and <0.1% sodium azide
Gene Alias HepPar-1
Host Species Mouse
Immunogen Formalin fixed failed human allograft liver that was mechanically disrupted actual price paid for the product
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.