Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HepPar-1 Mouse anti-Human, Clone: HepPar1, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14639
Description
Mouse monoclonal antibody raised against human HepPar-1.
- High specificity
- High reactivity
Specifications
HepPar-1 | |
Monoclonal | |
Unconjugated | |
Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
Protein A/G purification | |
RUO | |
Human | |
Liquid |
Immunofluorescence, Immunohistochemistry (PFA fixed) | |
HepPar1 | |
Mouse monoclonal antibody raised against human HepPar-1. | |
In 10 mM PBS (0.05% BSA and 0.05% azide). | |
Extract of a formalin-fixed, rejected-allograft of a human liver. | |
100 μg | |
Primary | |
Store at 4°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction